Learn More
Waters Corp XSelect HSS C18 Column, 100Å, 2.5 μm, 4.6 mm x 100 mm
Supplier: Waters Corp 186006147
These XSelect HSS C18 columns offer high-performance C18 chemistry with exceptional retention and peak shape. Utilizing proprietary High Strength Silica (HSS) Technology and ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns provide robust separation for HPLC assays and a seamless transition to UPLC.
- Exceptional retention and peak shape.
- Ultra-low dispersion hardware for robust separation and high throughput.
- High Strength Silica (HSS) Technology for unique selectivity.
- Withstands UPLC pressures up to 10000 psi (690 bar).
- Compatible with HPLC, UHPLC, and UPLC platforms.
- Trifunctional C18 ligand, USP L1, pH range 1 to 8.
- Endcapped, low silanol activity, spherical particles.
- Pore size: 100 Å; surface area: 230 m2/g; carbon load: 15%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.