Learn More
Waters Corp Waters Column, Cyano (CN), 2.5 μm, 2.1 mm, 150 mm
Catalog No. 50466456 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano column features a low coverage, unendcapped, and highly stable cyano phase, offering low hydrophobicity for rapid elution of hydrophobic analytes and alternate selectivity compared to traditional alkyl chain columns. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, providing exceptional separation performance, robustness, and throughput for HPLC assays, and facilitating a seamless transition to UPLC adoption.
  • Low coverage, unendcapped, highly stable cyano phase
  • Provides low hydrophobicity for the rapid elution of hydrophobic analytes
  • Offers alternate selectivity to traditional alkyl chain columns
  • Packed in ultra-low dispersion hardware
  • Enables exceptional separation performance, robustness and throughput for HPLC assays
  • Facilitates a seamless transition path toward UPLC adoption
Catalog No. 50-466-456 Supplier Waters Corp Supplier No. 186006748
The XSelect HSS Cyano column features a low coverage, unendcapped, and highly stable cyano phase, offering low hydrophobicity for rapid elution of hydrophobic analytes and alternate selectivity compared to traditional alkyl chain columns. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, providing exceptional separation performance, robustness, and throughput for HPLC assays, and facilitating a seamless transition to UPLC adoption.
  • Low coverage, unendcapped, highly stable cyano phase
  • Provides low hydrophobicity for the rapid elution of hydrophobic analytes
  • Offers alternate selectivity to traditional alkyl chain columns
  • Packed in ultra-low dispersion hardware
  • Enables exceptional separation performance, robustness and throughput for HPLC assays
  • Facilitates a seamless transition path toward UPLC adoption