Learn More
Waters Corp XSelect HSS Cyano (CN) XP Column, 100Å, 2.5 µm, 2.1 mm X 150 mm, 3/pk
Catalog No. 50465945 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that provides low hydrophobicity for the rapid elution of hydrophobic analytes as well as alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-465-945 Supplier Waters Corp Supplier No. 176002905
The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that provides low hydrophobicity for the rapid elution of hydrophobic analytes as well as alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.