Learn More
Waters Corp XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 2.5 µm, 2.1 mm X 150 mm, 3/pk
Catalog No. 50466498 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XSelect HSS CN columns packed with three different batches of sorbent. The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that provides low hydrophobicity for the rapid elution of hydrophobic analytes as well as alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-498 Supplier Waters Corp Supplier No. 186006806
This method validation kit consists of three XSelect HSS CN columns packed with three different batches of sorbent. The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that provides low hydrophobicity for the rapid elution of hydrophobic analytes as well as alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.