Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Method Validation Kit, Cyano (CN), 2.5 μm, 3 mm X 150 mm

Catalog No. 50466499 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect HSS Cyano (CN) column features a low coverage, unendcapped, highly stable cyano phase, delivering low hydrophobicity for rapid elution of hydrophobic analytes and alternate selectivity. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns ensure exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC adoption.

  • Provides low hydrophobicity for rapid elution of hydrophobic analytes and alternate selectivity.
  • Kit includes three columns packed with three different batches of sorbent for method validation.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Facilitates seamless transition to UPLC adoption.
  • Particle size: 2.5 μm.
  • Pore size: 100 Å.
  • Inner diameter: 3 mm; Length: 150 mm.
  • pH range: 2 to 8 pH.
  • Maximum pressure: 18000 psi (1240 Bar).

Catalog No. 50-466-499 Supplier Waters Corp Supplier No. 186006807
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect HSS Cyano (CN) column features a low coverage, unendcapped, highly stable cyano phase, delivering low hydrophobicity for rapid elution of hydrophobic analytes and alternate selectivity. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns ensure exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC adoption.

  • Provides low hydrophobicity for rapid elution of hydrophobic analytes and alternate selectivity.
  • Kit includes three columns packed with three different batches of sorbent for method validation.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Facilitates seamless transition to UPLC adoption.
  • Particle size: 2.5 μm.
  • Pore size: 100 Å.
  • Inner diameter: 3 mm; Length: 150 mm.
  • pH range: 2 to 8 pH.
  • Maximum pressure: 18000 psi (1240 Bar).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.