Learn More
Waters Corp XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 2.5 µm, 4.6 mm X 150 mm, 3/pk
Catalog No. 50466500 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
coverage, unendcapped, highly stable cyano phase that provides low hydrophobicity for the rapid elution of hydrophobic analytes as well as alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-500 Supplier Waters Corp Supplier No. 186006808
coverage, unendcapped, highly stable cyano phase that provides low hydrophobicity for the rapid elution of hydrophobic analytes as well as alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.