Learn More
Waters Corp Waters XSelect HSS Cyano (CN) Column, 2.5 μm, 100 Å, 4.6 mm × 150 mm
Supplier:  Waters Corp 186006750
Encompass_Preferred
The XSelect HSS Cyano column features a low coverage, unendcapped, and highly stable cyano phase, optimized for rapid elution of hydrophobic analytes and alternate selectivity. Built with HSS (High Strength Silica) technology, these 2.5 μm eXtended Performance [XP] columns provide exceptional separation, robustness, and extended lifetimes for both HPLC and UPLC applications, supporting both reversed-phase and normal-phase chromatography.
  • Low coverage, unendcapped, and highly stable cyano phase.
  • Delivers low hydrophobicity for rapid elution of hydrophobic analytes and offers alternate selectivity.
  • Enhances chromatographic performance and polar molecule retention with HSS technology.
  • Designed for UPLC pressures and provides long column lifetimes at low pH.
  • Cost-effective due to mild retention, allowing less organic solvent and faster analysis.
Catalog No. 50-466-458