Learn More
Waters Corp Column, HSS CN, 3.5 μm, 3.0 x 150 mm
Catalog No. 50179389 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS CN Column is an HPLC column designed for a wide range of chromatographic separations. It features HSS CN chemistry with a 3.5 μm particle size and 100 Å pore diameter, allowing for both normal-phase and reversed-phase separations.
  • Designed for the widest range of chromatographic separations.
  • Can be successfully used for normal-phase and reversed-phase separations.
  • Offers diverse analyte selectivity.
  • Provides high analyte loading and operational stability.
  • Full scalability for UPLC, HPLC, and preparative separation platforms.
  • Seamlessly scalable and possesses the same chemical and physical characteristics as the 1.8 μm HSS particle.
  • Offers superior peak shape for basic compounds, low column bleed, excellent batch-to-batch reproducibility, ultra-efficiencies, and seamless transferability between different particle sizes and column formats.
Catalog No. 50-179-389 Supplier Waters Corp Supplier No. 186005916
The XSelect HSS CN Column is an HPLC column designed for a wide range of chromatographic separations. It features HSS CN chemistry with a 3.5 μm particle size and 100 Å pore diameter, allowing for both normal-phase and reversed-phase separations.
  • Designed for the widest range of chromatographic separations.
  • Can be successfully used for normal-phase and reversed-phase separations.
  • Offers diverse analyte selectivity.
  • Provides high analyte loading and operational stability.
  • Full scalability for UPLC, HPLC, and preparative separation platforms.
  • Seamlessly scalable and possesses the same chemical and physical characteristics as the 1.8 μm HSS particle.
  • Offers superior peak shape for basic compounds, low column bleed, excellent batch-to-batch reproducibility, ultra-efficiencies, and seamless transferability between different particle sizes and column formats.