Learn More
Waters Corp Waters HPLC Column, Cyano (CN), 3.5 μm, 4.6 mm X 250 mm
Supplier:  Waters Corp 186005923
Encompass_Preferred
The XSelect HSS CN chemistry is an ultra-performance general purpose propyl cyano bonded phase, offering superior peak shape for bases, increased retention, and extended column lifetimes at low pH. These HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
  • Efficient for separating polar compounds.
  • 100 Å pore size retains polar compounds.
  • 3.5 μm particle size separates small polar compounds.
  • Cyano (CN) bonded phase for polar compound separation (e.g., amino acids, peptides, carbohydrates).
  • Wide pH range of 2–8 with low pH stability for diverse applications like peptide mapping.
  • Ultra-low column bleed.
  • Manufactured in cGMP, ISO 9001:2000 certified plant; each column individually tested.
Catalog No. 50-179-396