Learn More
Waters Corp HPLC Column, XSelect HSS Cyano, 3.5 μm, 4.6 x 75 mm
Supplier:  Waters Corp 186005920
Encompass_Preferred
This analytical column is part of the XSelect HPLC Column family, offering exceptional selectivity for a wide range of chromatographic separations. Featuring an XSelect HSS Cyano phase, it combines specific ligands with proprietary HSS Technologies, making it suitable for both normal-phase and reversed-phase separations.
  • Can be used for normal-phase and reversed-phase separations.
  • Provides uniquely selective separations with proprietary HSS Technologies.
  • Delivers high analyte loading and operational stability.
  • Offers full scalability for UPLC™, HPLC, and preparative separation platforms.
  • Provides excellent batch-to-batch reproducibility and superior peak shape.
  • Seamless transferability between different particle sizes and formats.
  • HSS particle offers increased polar compound retention and high mechanical strength.
Catalog No. 50-179-393