Learn More
Waters Corp WATERS CORPORATION Intelligent Speed Column, Cyano, 3.5 μm, 2.1 × 20 mm
Catalog No. 50179378 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This intelligent speed column features a propyl cyano bonded phase, offering superior peak shape for bases, increased retention, and extended column lifetimes at low pH. It is fully scalable and transferable to UPLC separations, designed to aid method development. The column includes high performance surface technology to minimize unwanted analyte-surface interactions.
  • Ultra-performance propyl cyano bonded phase.
  • Superior peak shape for bases and increased retention of polar compounds.
  • Extended column lifetimes and excellent stability at low pH.
  • Fully scalable and transferable to UPLC separations.
  • High performance surface technology prevents analyte-surface interactions.
  • Flexible tool for methods development, ensuring stable and reproducible separations.
Catalog No. 50-179-378 Supplier Waters Corp Supplier No. 186005905
This intelligent speed column features a propyl cyano bonded phase, offering superior peak shape for bases, increased retention, and extended column lifetimes at low pH. It is fully scalable and transferable to UPLC separations, designed to aid method development. The column includes high performance surface technology to minimize unwanted analyte-surface interactions.
  • Ultra-performance propyl cyano bonded phase.
  • Superior peak shape for bases and increased retention of polar compounds.
  • Extended column lifetimes and excellent stability at low pH.
  • Fully scalable and transferable to UPLC separations.
  • High performance surface technology prevents analyte-surface interactions.
  • Flexible tool for methods development, ensuring stable and reproducible separations.