Learn More
Waters Corp XSelect HSS Cyano (CN) Column, 100Å, 5 μm, 3 mm x 250 mm
Catalog No. 50179410 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS CN chemistry is a general-purpose propyl cyano bonded phase designed for ultra-performance. It offers superior peak shape for bases, increased retention, and extended column lifetimes at low pH. These HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
  • Provides superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to ACQUITY UPLC separations
Catalog No. 50-179-410 Supplier Waters Corp Supplier No. 186005939
The XSelect HSS CN chemistry is a general-purpose propyl cyano bonded phase designed for ultra-performance. It offers superior peak shape for bases, increased retention, and extended column lifetimes at low pH. These HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
  • Provides superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to ACQUITY UPLC separations