Learn More
Waters Corp Waters HPLC Column, HSS Cyano, 5 μm, 100 Å, 4.6 x 30 mm
Supplier:  Waters Corp 186005941
Encompass_Preferred
The HSS Cyano (CN) column is a versatile HPLC column, leveraging proprietary High Strength Silica (HSS) Technology. It offers unique selectivity, high analyte loading, and robust operational stability across UPLC™, HPLC, and preparative systems. This adaptable phase facilitates successful normal-phase and reversed-phase separations for complex samples.
  • Diverse selectivity for normal-phase and reversed-phase separation.
  • Increased retention for polar compounds due to higher silanophilicity.
  • Mechanical strength to withstand high pressures, ensuring reliable performance.
  • Scalable particle sizes (2.5, 3.5, 5 μm) ensure consistent method transferability.
  • Improved resolution of complex samples for isolation and quantification.
  • Operational stability and excellent batch-to-batch reproducibility.
Catalog No. 50-179-412