Learn More
Waters Corp HPLC Column, Cyano, 5 μm, 4.6 mm ID, 50 mm Length
Supplier:  Waters Corp 186005942
Encompass_Preferred
The XSelect HSS Cyano HPLC Column is designed for diverse analyte selectivity and can be used for both normal-phase and reversed-phase separations. It offers increased polar compound retention and possesses mechanical strength for UPLC pressures, ensuring reproducibility and scalability.
  • High Strength Silica Technology
  • Monofunctional Cyano-Propyl phase
  • Increased polar compound retention compared to ethylene-bridge-containing BEH and CSH particles
  • Mechanical strength to tolerate UPLC pressures
  • Seamlessly scalable with consistent chemical and physical characteristics
  • Designed to provide diverse analyte selectivity
  • Contributes to unmatched reproducibility and selectivity
  • pH range of 2–8
  • Temperature limits of 45 °C for both low and high pH conditions
Catalog No. 50-179-413