Learn More
Waters Corp VanGuard Cartridge, XSelect HSS Cyano (CN), 3.5 μm, 2.1 x 5 mm
Catalog No. 502391298 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano (CN) VanGuard Cartridge is designed to extend the lifetime of analytical columns while maintaining chromatographic performance. With dimensions of 2.1 x 5 mm and a 3.5 μm particle size, this cartridge provides physical and chemical protection for analytical columns.
  • Removes particulates and chemical contamination.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Provides cost-effective protection for analytical columns.
Catalog No. 50-239-1298 Supplier Waters Corp Supplier No. 186007860
The XSelect HSS Cyano (CN) VanGuard Cartridge is designed to extend the lifetime of analytical columns while maintaining chromatographic performance. With dimensions of 2.1 x 5 mm and a 3.5 μm particle size, this cartridge provides physical and chemical protection for analytical columns.
  • Removes particulates and chemical contamination.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Provides cost-effective protection for analytical columns.