Learn More
Waters Corp XSelect HSS Cyano (CN) VanGuard Cartridge, 100Å, 5 μm, 3.9 mm X 5 mm
Supplier:  Waters Corp 186007865
Encompass_Preferred
This guard cartridge works in conjunction with an analytical column by attaching to it. It protects the column from harm by removing all chemical and particulate matter from the mobile phase, enhancing the column's lifetime and ensuring outstanding results throughout its service.
  • Protects the column from harm.
  • Removes chemical and particulate matter from the mobile phase.
  • Enhances the column's lifetime.
  • Ensures the column continues to deliver outstanding results.
Catalog No. 50-239-1933