Learn More
Waters Corp WATERS CORPORATION Method Validation Kit, XSelect HSS CN, 2.1 x 100 mm, 3.5 μm
Catalog No. 50179421 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano Method Validation Kit is designed for chromatographic challenges requiring diverse selectivity. It contains three HSS CN columns, each with a 2.1 x 100 mm dimension and 3.5 μm particle size, from different batches to ensure reproducibility and scalability for method validation.
  • Provides exceptional selectivity for successful separations.
  • Combines carefully chosen ligands with proprietary High Strength Silica (HSS) technology.
  • Designed to provide diverse analyte selectivity for separating closely eluting sample constituents.
  • High Strength Silica (HSS) particles offer increased polar compound retention and mechanical strength to tolerate UPLC pressures.
  • Effectively utilized for both normal-phase and reversed-phase separations.
  • Seamlessly scalable across different particle sizes with consistent chemical and physical characteristics.
Catalog No. 50-179-421 Supplier Waters Corp Supplier No. 186005950
The XSelect HSS Cyano Method Validation Kit is designed for chromatographic challenges requiring diverse selectivity. It contains three HSS CN columns, each with a 2.1 x 100 mm dimension and 3.5 μm particle size, from different batches to ensure reproducibility and scalability for method validation.
  • Provides exceptional selectivity for successful separations.
  • Combines carefully chosen ligands with proprietary High Strength Silica (HSS) technology.
  • Designed to provide diverse analyte selectivity for separating closely eluting sample constituents.
  • High Strength Silica (HSS) particles offer increased polar compound retention and mechanical strength to tolerate UPLC pressures.
  • Effectively utilized for both normal-phase and reversed-phase separations.
  • Seamlessly scalable across different particle sizes with consistent chemical and physical characteristics.