Learn More
Waters Corp WATERS CORPORATION Method Validation Kit, Cyano (CN), 5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186005955
Encompass_Preferred
The XSelect HSS Cyano (CN) Method Validation Kit features an ultra-performance propyl cyano bonded phase for superior peak shape and increased retention. It includes three identical columns from different lots for robust method validation, offering extended column lifetimes at low pH and excellent stability.
  • Fully scalable and transferable to ACQUITY UPLC separations
  • Low-coverage, highly stable cyano phase
  • Wide and usable pH range
  • Highly resistant to extreme backpressures
  • Utilizes High-Strength Silica (HSS) base particle
Catalog No. 50-179-426