Learn More
Waters Corp XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 2.5 µm, 2.1 mm X 100 mm, 3/pk
Supplier:  Waters Corp 186006276
Encompass_Preferred
This method validation kit consists of three batches of XSelect HSS Cyano columns packed with three different batches of sorbent
Catalog No. 50-179-629