Learn More
Waters Corp Waters Corporation XSelect HSS Cyano (CN) XP Column, 100Å, 2.5 μm, 3 mm Inner Diameter, 50 mm Length
Catalog No. 50179314 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano column is a stable cyano phase, offering low hydrophobicity for the elution of hydrophobic analytes. These 2.5 μm XP columns provide exceptional separation performance for HPLC assays and facilitate a seamless transition to UPLC.
  • Stable and reproducible cyano chemistry, suitable for rapid reproduction of analyses to validate data.
  • Incorporates MaxPeak High-Performance Surface technology to prevent unwanted analyte-surface interactions.
  • Offers alternate selectivity and increased retention of polar compounds, as well as high efficiency under demanding conditions.
  • pH range of 2 to 8.
  • Can withstand pressures up to 18,000 psi.
  • High-Strength Silica (HSS) base particle allows handling various analytes without compromising performance.
Catalog No. 50-179-314 Supplier Waters Corp Supplier No. 176002655
Add to Cart
Add to Cart
The XSelect HSS Cyano column is a stable cyano phase, offering low hydrophobicity for the elution of hydrophobic analytes. These 2.5 μm XP columns provide exceptional separation performance for HPLC assays and facilitate a seamless transition to UPLC.
  • Stable and reproducible cyano chemistry, suitable for rapid reproduction of analyses to validate data.
  • Incorporates MaxPeak High-Performance Surface technology to prevent unwanted analyte-surface interactions.
  • Offers alternate selectivity and increased retention of polar compounds, as well as high efficiency under demanding conditions.
  • pH range of 2 to 8.
  • Can withstand pressures up to 18,000 psi.
  • High-Strength Silica (HSS) base particle allows handling various analytes without compromising performance.