Learn More
Waters Corp Waters XSelect HSS Cyano Column, 100 Å, 2.5 μm, 3 mm x 50 mm
Catalog No. 50179630 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three batches of XSelect HSS Cyano columns packed with three different batches of sorbent.
  • Cyano (CN) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8 pH
  • Max pressure: 18000 psi (1240 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting
  • 100 Å pore size
  • Column format
  • Surface area of 230
  • UPLC and UHPLC system compatible
  • HSS particle technology
  • USP classification L10
  • 3 mm inner diameter
  • 50 mm length
  • 5% carbon load
  • 3 units per package
  • Digital Certificates of Analysis and eCords
Catalog No. 50-179-630 Supplier Waters Corp Supplier No. 186006277
This method validation kit consists of three batches of XSelect HSS Cyano columns packed with three different batches of sorbent.
  • Cyano (CN) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8 pH
  • Max pressure: 18000 psi (1240 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting
  • 100 Å pore size
  • Column format
  • Surface area of 230
  • UPLC and UHPLC system compatible
  • HSS particle technology
  • USP classification L10
  • 3 mm inner diameter
  • 50 mm length
  • 5% carbon load
  • 3 units per package
  • Digital Certificates of Analysis and eCords