Learn More
Waters Corp Waters Column, Cyano (CN), 2.5 μm, 4.6 x 100 mm
Supplier:  Waters Corp 186006195
Encompass_Preferred
This cyano column provides low hydrophobicity for rapid elution of hydrophobic analytes and offers alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, with a seamless transition path toward UPLC adoption.
  • Provides superior peak shape for bases and extended column life at low pH.
  • Suitable for both reversed-phase and normal-phase chromatography.
  • Fully scalable and convertible to ACQUITY UPLC separations.
  • Utilizes HSS (High Strength Silica) technology for optimal separation, greater polar molecule retention, and dramatically different selectivity.
  • Offers mechanical strength required for UPLC pressures.
  • Moderate retention properties allow for less organic solvent and faster chromatographic analysis.
Catalog No. 50-201-576