Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, Cyano (CN), 2.5 μm, 100 Å, 3 mm x 30 mm

Catalog No. 50179313 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect HSS Cyano column offers a low coverage, unendcapped, and highly stable cyano phase, providing low hydrophobicity for rapid elution of hydrophobic analytes and alternative selectivity. Featuring ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and throughput for HPLC assays, facilitating a smooth transition to UPLC.

  • Cyano (CN) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • Particle size of 2.5 μm
  • Pore size of 100 Å
  • Inner diameter of 3 mm
  • Length of 30 mm
  • Compatible with UPLC and UHPLC systems

Catalog No. 50-179-313 Supplier Waters Corp Supplier No. 176002654
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect HSS Cyano column offers a low coverage, unendcapped, and highly stable cyano phase, providing low hydrophobicity for rapid elution of hydrophobic analytes and alternative selectivity. Featuring ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and throughput for HPLC assays, facilitating a smooth transition to UPLC.

  • Cyano (CN) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • Particle size of 2.5 μm
  • Pore size of 100 Å
  • Inner diameter of 3 mm
  • Length of 30 mm
  • Compatible with UPLC and UHPLC systems

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.