Learn More
Waters Corp Waters XSelect HSS Cyano Column, 100Å, 2.5 μm, 2.1 mm × 50 mm
Catalog No. 50179569 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XSelect HSS Cyano Column, 100Å, 2.5 μm, 2.1 mm × 50 mm
Catalog No. 50-179-569 Supplier Waters Corp Supplier No. 186006185
Waters XSelect HSS Cyano Column, 100Å, 2.5 μm, 2.1 mm × 50 mm