Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, Cyano (CN), 100Å, 2.5 μm, 2.1 mm X 30 mm

Catalog No. 50179309 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that offers low hydrophobicity for the rapid elution of hydrophobic analytes, and provides an alternate selectivity compared to traditional alkyl chain columns. When packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a smooth transition path towards UPLC adoption.

  • Chemistry: Cyano (CN)
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8 pH
  • Maximum pressure: 18000 psi (1240 bar)
  • Particle size: 2.5 μm
  • Pore size: 100Å
  • Inner diameter: 2.1 mm
  • Length: 30 mm
  • System: UPLC, UHPLC
  • Units per package: 3

Catalog No. 50-179-309 Supplier Waters Corp Supplier No. 176002650
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that offers low hydrophobicity for the rapid elution of hydrophobic analytes, and provides an alternate selectivity compared to traditional alkyl chain columns. When packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a smooth transition path towards UPLC adoption.

  • Chemistry: Cyano (CN)
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8 pH
  • Maximum pressure: 18000 psi (1240 bar)
  • Particle size: 2.5 μm
  • Pore size: 100Å
  • Inner diameter: 2.1 mm
  • Length: 30 mm
  • System: UPLC, UHPLC
  • Units per package: 3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.