Learn More
Waters Corp Waters Column, Cyano (CN), 2.5 μm, 100 Å, 3 mm x 75 mm
Supplier:  Waters Corp 176002656
Encompass_Preferred
The XSelect HSS Cyano column provides low hydrophobicity and alternate selectivity for rapid elution of hydrophobic analytes. Featuring HSS base particles and MaxPeak High-Performance Surface Technology, these 2.5 μm XP columns offer exceptional HPLC assay performance and enable seamless UPLC/UHPLC transitions.
  • Low coverage, unendcapped, highly stable cyano phase
  • Packed in ultra-low dispersion hardware for exceptional separation and throughput
  • Offers robustness for HPLC assays
  • Features a pH range of 2 to 8 pH
  • Supports a maximum pressure of 18,000 PSI
  • Compatible with VanGuard Cartridges for protection
Catalog No. 50-179-315