Learn More
Waters Corp Waters Column, Cyano (CN), 2.5 μm, 100Å, 3 × 75 mm
Catalog No. 50179574 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano column is a low-coverage, unendcapped, highly stable cyano phase offering low hydrophobicity for rapid hydrophobic analyte elution and alternative selectivity. These 2.5 μm XP columns utilize ultra-low dispersion hardware for exceptional separation performance, robustness, and throughput in HPLC assays, also facilitating seamless UPLC adoption.
  • Cyano (CN) chemistry
  • Reversed phase separation
  • pH 2 to 8
  • Maximum pressure: 18000 psi (1240 Bar)
  • Unendcapped
  • Spherical particle shape
  • 2.5 μm particle size
  • 100 Å pore size
  • UPLC and UHPLC compatible
  • HSS particle technology
  • L10 USP classification
  • 3 mm ID x 75 mm length
  • 5% carbon load
  • Single unit package
  • Parker-style endfitting
Catalog No. 50-179-574 Supplier Waters Corp Supplier No. 186006190
The XSelect HSS Cyano column is a low-coverage, unendcapped, highly stable cyano phase offering low hydrophobicity for rapid hydrophobic analyte elution and alternative selectivity. These 2.5 μm XP columns utilize ultra-low dispersion hardware for exceptional separation performance, robustness, and throughput in HPLC assays, also facilitating seamless UPLC adoption.
  • Cyano (CN) chemistry
  • Reversed phase separation
  • pH 2 to 8
  • Maximum pressure: 18000 psi (1240 Bar)
  • Unendcapped
  • Spherical particle shape
  • 2.5 μm particle size
  • 100 Å pore size
  • UPLC and UHPLC compatible
  • HSS particle technology
  • L10 USP classification
  • 3 mm ID x 75 mm length
  • 5% carbon load
  • Single unit package
  • Parker-style endfitting