Learn More
Waters Corp XSelect Column, Cyano (CN), 2.5 μm, 100 Å, 2.1 mm x 75 mm
Catalog No. 50179570 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that offers low hydrophobicity for the rapid elution of hydrophobic analytes. It also provides an alternate selectivity compared to traditional alkyl chain columns. The 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays, and facilitating a seamless transition towards UPLC adoption.
  • Low coverage, unendcapped, highly stable cyano phase.
  • Low hydrophobicity for rapid elution of hydrophobic analytes.
  • Alternate selectivity to traditional alkyl chain columns.
  • Packed in ultra-low dispersion hardware (2.5 μm XP columns).
  • Exceptional separation performance, robustness, and throughput for HPLC.
  • Seamless transition path toward UPLC adoption.
Catalog No. 50-179-570 Supplier Waters Corp Supplier No. 186006186
The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase that offers low hydrophobicity for the rapid elution of hydrophobic analytes. It also provides an alternate selectivity compared to traditional alkyl chain columns. The 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays, and facilitating a seamless transition towards UPLC adoption.
  • Low coverage, unendcapped, highly stable cyano phase.
  • Low hydrophobicity for rapid elution of hydrophobic analytes.
  • Alternate selectivity to traditional alkyl chain columns.
  • Packed in ultra-low dispersion hardware (2.5 μm XP columns).
  • Exceptional separation performance, robustness, and throughput for HPLC.
  • Seamless transition path toward UPLC adoption.