Learn More
Waters Corp XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 2.5 µm, 2.1 mm X 50 mm, 3/pk
Catalog No. 50179628 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three batches of XSelect HSS Cyano columns packed with three different batches of sorbent
Catalog No. 50-179-628 Supplier Waters Corp Supplier No. 186006275
This method validation kit consists of three batches of XSelect HSS Cyano columns packed with three different batches of sorbent