Learn More
Waters Corp WATERS CORPORATION XSelect HSS Cyano Column, 100Å, 2.5 μm, 4.6 mm x 50 mm
Supplier: Waters Corp 186006193
The XSelect HSS Cyano column features a low coverage, unendcapped, highly stable cyano phase, ideal for rapid elution of hydrophobic analytes and offering alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns ensure exceptional separation performance, robustness, and throughput for HPLC assays, enabling seamless UPLC transition.
- Chemistry is Cyano (CN)
- Separation mode is Reversed Phase
- Particle substrate is Silica
- pH range is 2 pH to 8 pH
- Maximum pressure is 10000 psi (690 Bar)
- Not endcapped
- High Silanol activity
- Particle shape is Spherical
- Particle size is 2.5 μm
- Endfitting type is Parker-style
- Pore size is 100 Å
- Compatible with UPLC and UHPLC systems
- Inner diameter is 4.6 mm
- Length is 50 mm
- Carbon load is 5 %
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.