Learn More
Waters Corp WATERS CORPORATION XSelect HSS Cyano (CN) Column, 100Å, 2.5 μm, 2.1 mm X 50 mm
Catalog No. 50179310 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS Cyano (CN) Column features a 100Å pore size and 2.5 μm particle size, offering precise separation for various analytical applications. This column measures 2.1 mm X 50 mm and is supplied as a 3-pack, suitable for laboratories requiring multiple columns with consistent performance.
  • 100Å pore size for optimal separation.
  • 2.5 μm particle size ensures high resolution.
  • Dimensions of 2.1 mm X 50 mm for versatile use.
  • Cyano (CN) chemistry for unique selectivity.
  • High strength silica (HSS) for extended column lifetime.
  • Supplied in a convenient 3-pack.
Catalog No. 50-179-310 Supplier Waters Corp Supplier No. 176002651
The XSelect HSS Cyano (CN) Column features a 100Å pore size and 2.5 μm particle size, offering precise separation for various analytical applications. This column measures 2.1 mm X 50 mm and is supplied as a 3-pack, suitable for laboratories requiring multiple columns with consistent performance.
  • 100Å pore size for optimal separation.
  • 2.5 μm particle size ensures high resolution.
  • Dimensions of 2.1 mm X 50 mm for versatile use.
  • Cyano (CN) chemistry for unique selectivity.
  • High strength silica (HSS) for extended column lifetime.
  • Supplied in a convenient 3-pack.