Learn More
Waters Corp Waters Column, Cyano (CN), 2.5 μm, 2.1 mm x 30 mm
Supplier: Waters Corp 186006184
The XSelect HSS Cyano column is a low coverage, unendcapped, highly stable cyano phase, offering low hydrophobicity for rapid elution of hydrophobic analytes. It provides alternate selectivity to traditional alkyl chain columns. Packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns ensure exceptional separation, robustness, and throughput for HPLC assays, supporting UPLC adoption.
- Cyano (CN) chemistry for reversed phase separation.
- Silica particle substrate with spherical shape.
- pH range of 2 to 8.
- Maximum pressure of 18000 psi (1240 bar).
- Unendcapped phase with high silanol activity.
- Particle size: 2.5 μm; Pore size: 100 Å.
- Suitable for UPLC and UHPLC systems with HSS particle technology.
- Dimensions: 2.1 mm inner diameter, 30 mm length.
- 5% carbon load.
- Packaged with one column per pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.