Learn More
Waters Corp Waters XSelect HSS PFP Method Validation Kit, 2.5 μm, 100 Å
Supplier:  Waters Corp 186006816
Encompass_Preferred
This method validation kit provides three high-performance columns, each with a unique sorbent batch. Utilizing a low coverage, unendcapped pentafluorophenyl phase, these columns enable superior separation of complex mixtures and offer distinct selectivity from conventional alkyl chain columns.
  • Includes three columns with different sorbent batches for validation
  • Low coverage, unendcapped PFP phase for unique selectivity
  • Exceptional separation of closely related compounds
  • 2.5 μm XP particles in ultra-low dispersion hardware
  • High performance, robustness, and throughput for HPLC/UPLC
  • PFP/Fluoro-Phenyl chemistry, Reversed Phase separation
  • Silica, spherical particles; 100 Å pore size
  • 2.1 mm inner diameter, 100 mm length
  • pH range: 2 to 8; Max pressure: 18000 psi (1240 Bar)
Catalog No. 50-466-505