Learn More
Waters Corp Waters XSelect HSS PFP Method Validation Kit, 2.5 μm, 2.1 mm, 3 pk
Catalog No. 50466495 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns, each packed with a different batch of sorbent. The columns feature a low coverage, unendcapped pentafluorophenyl phase, offering exceptional separation of closely related compounds and alternative selectivity compared to traditional alkyl chain columns. They are packed in ultra-low dispersion hardware, and their 2.5 μm eXtended Performance [XP] designation indicates they are designed for exceptional separation performance, robustness, and throughput in HPLC assays, while also facilitating a seamless transition to UPLC adoption.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation mode
  • 2.5 μm particle size
  • 2.1 mm inner diameter
  • 150 mm length
  • Pore size of 100 Å
  • pH range of 2 to 8
  • Maximum pressure of 18000 psi (1240 bar)
  • Three units per package
Catalog No. 50-466-495 Supplier Waters Corp Supplier No. 186006803
This method validation kit includes three columns, each packed with a different batch of sorbent. The columns feature a low coverage, unendcapped pentafluorophenyl phase, offering exceptional separation of closely related compounds and alternative selectivity compared to traditional alkyl chain columns. They are packed in ultra-low dispersion hardware, and their 2.5 μm eXtended Performance [XP] designation indicates they are designed for exceptional separation performance, robustness, and throughput in HPLC assays, while also facilitating a seamless transition to UPLC adoption.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation mode
  • 2.5 μm particle size
  • 2.1 mm inner diameter
  • 150 mm length
  • Pore size of 100 Å
  • pH range of 2 to 8
  • Maximum pressure of 18000 psi (1240 bar)
  • Three units per package