Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Method Validation Kit, PFP/Fluoro-Phenyl, 2.5 μm, 100Å, 2.1 mm X 50 mm

Catalog No. 50466504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit contains three columns for method validation, featuring PFP/Fluoro-Phenyl chemistry and 2.5 μm XP particle technology. Designed for HPLC assays, they offer superior separation, robustness, and enable UPLC transition.

  • Includes three columns with varied sorbent batches for method validation.
  • PFP/Fluoro-Phenyl chemistry provides unique selectivity for closely related compounds.
  • Ultra-low dispersion hardware with 2.5 μm XP particle technology.
  • Optimized for HPLC assays, offering high performance, robustness, and throughput.
  • Facilitates seamless transition to UPLC.
  • Specifications: Reversed phase, Silica substrate, Spherical particles.
  • Dimensions: 2.1 mm ID, 50 mm length, 100 Å pore size.
  • Operating range: pH 2 to 8, Maximum pressure 18000 psi (1240 Bar).

Catalog No. 50-466-504 Supplier Waters Corp Supplier No. 186006815
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit contains three columns for method validation, featuring PFP/Fluoro-Phenyl chemistry and 2.5 μm XP particle technology. Designed for HPLC assays, they offer superior separation, robustness, and enable UPLC transition.

  • Includes three columns with varied sorbent batches for method validation.
  • PFP/Fluoro-Phenyl chemistry provides unique selectivity for closely related compounds.
  • Ultra-low dispersion hardware with 2.5 μm XP particle technology.
  • Optimized for HPLC assays, offering high performance, robustness, and throughput.
  • Facilitates seamless transition to UPLC.
  • Specifications: Reversed phase, Silica substrate, Spherical particles.
  • Dimensions: 2.1 mm ID, 50 mm length, 100 Å pore size.
  • Operating range: pH 2 to 8, Maximum pressure 18000 psi (1240 Bar).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.