Learn More
Waters Corp Waters Corporation Method Validation Kit, PFP, 2.5 μm, 100 Å, 3 x 150 mm
Catalog No. 50466496 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns, each packed with a different sorbent batch. These columns feature a low coverage, unendcapped pentafluorophenyl phase, offering exceptional separation of closely related compounds and alternative selectivity. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance (XP) columns deliver excellent separation performance, robustness, and throughput for HPLC assays, supporting UPLC adoption.
  • Consists of three columns packed with three different batches of sorbent.
  • Features a low coverage, unendcapped pentafluorophenyl phase.
  • Provides exceptional separation of closely related compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance (XP) columns enable exceptional separation, robustness, and throughput for HPLC assays, supporting UPLC adoption.
Catalog No. 50-466-496 Supplier Waters Corp Supplier No. 186006804
This method validation kit includes three columns, each packed with a different sorbent batch. These columns feature a low coverage, unendcapped pentafluorophenyl phase, offering exceptional separation of closely related compounds and alternative selectivity. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance (XP) columns deliver excellent separation performance, robustness, and throughput for HPLC assays, supporting UPLC adoption.
  • Consists of three columns packed with three different batches of sorbent.
  • Features a low coverage, unendcapped pentafluorophenyl phase.
  • Provides exceptional separation of closely related compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance (XP) columns enable exceptional separation, robustness, and throughput for HPLC assays, supporting UPLC adoption.