Learn More
Waters Corp Waters Method Validation Kit, PFP, 2.5 μm, 3 mm × 100 mm, 3 pk
Catalog No. 50466507 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three HSS PFP columns with varying sorbent batches. These columns offer low coverage, unendcapped pentafluorophenyl phase for exceptional separation of related compounds and alternate selectivity. With ultra-low dispersion hardware and 2.5 μm XP columns, it ensures robust, high-throughput HPLC assays and facilitates UPLC transition.
  • Low coverage, unendcapped pentafluorophenyl (PFP) phase
  • Exceptional separation of closely related compounds
  • Alternate selectivity to traditional alkyl chain columns
  • Packed in ultra-low dispersion hardware
  • 2.5 μm eXtended Performance [XP] columns
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays
  • Seamless transition path toward UPLC adoption
  • Method validation kit consisting of three columns with different sorbent batches
Catalog No. 50-466-507 Supplier Waters Corp Supplier No. 186006818
This method validation kit includes three HSS PFP columns with varying sorbent batches. These columns offer low coverage, unendcapped pentafluorophenyl phase for exceptional separation of related compounds and alternate selectivity. With ultra-low dispersion hardware and 2.5 μm XP columns, it ensures robust, high-throughput HPLC assays and facilitates UPLC transition.
  • Low coverage, unendcapped pentafluorophenyl (PFP) phase
  • Exceptional separation of closely related compounds
  • Alternate selectivity to traditional alkyl chain columns
  • Packed in ultra-low dispersion hardware
  • 2.5 μm eXtended Performance [XP] columns
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays
  • Seamless transition path toward UPLC adoption
  • Method validation kit consisting of three columns with different sorbent batches