Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation XSelect HSS PFP Method Validation Kit, 2.5 μm

Catalog No. 50466497 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit features three columns packed with different batches of sorbent, utilizing a low coverage, unendcapped pentafluorophenyl phase. It offers exceptional separation and alternate selectivity to traditional alkyl chain columns, providing excellent performance for HPLC assays and a seamless transition to UPLC.

  • Consists of three columns for method validation.
  • Packed in ultra-low dispersion hardware.
  • eXtended Performance (XP) columns with 2.5 μm particle size.
  • Wide pH range: 2 to 8.
  • Maximum pressure: 10000 psi (690 Bar).
  • High silanol activity.
  • Spherical particle shape.
  • Parker-style endfitting.
  • Pore size: 100 Å.
  • Surface area: 230.
  • Compatible with UPLC and UHPLC systems.
  • USP classification: L43.
  • Carbon load: 7%.
  • Inner diameter: 4.6 mm.
  • Length: 150 mm.
  • Units per package: 3.

Catalog No. 50-466-497 Supplier Waters Corp Supplier No. 186006805
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit features three columns packed with different batches of sorbent, utilizing a low coverage, unendcapped pentafluorophenyl phase. It offers exceptional separation and alternate selectivity to traditional alkyl chain columns, providing excellent performance for HPLC assays and a seamless transition to UPLC.

  • Consists of three columns for method validation.
  • Packed in ultra-low dispersion hardware.
  • eXtended Performance (XP) columns with 2.5 μm particle size.
  • Wide pH range: 2 to 8.
  • Maximum pressure: 10000 psi (690 Bar).
  • High silanol activity.
  • Spherical particle shape.
  • Parker-style endfitting.
  • Pore size: 100 Å.
  • Surface area: 230.
  • Compatible with UPLC and UHPLC systems.
  • USP classification: L43.
  • Carbon load: 7%.
  • Inner diameter: 4.6 mm.
  • Length: 150 mm.
  • Units per package: 3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.