Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, PFP Fluoro-Phenyl, 2.5 μm, 3 mm x 150 mm

Catalog No. 50466454 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

  • Low coverage, unendcapped pentafluorophenyl phase
  • Offers exceptional separation of closely related compounds
  • Provides alternate selectivity to traditional alkyl chain columns
  • Packed in ultra-low dispersion hardware for eXtended Performance (XP)
  • Enables robust separation performance and high throughput for HPLC assays
  • Facilitates seamless transition to UPLC adoption
  • PFP/Fluoro-Phenyl chemistry, reversed phase separation mode
  • Silica particle substrate with spherical shape
  • pH range of 2 to 8
  • Maximum pressure of 18000 psi (1240 Bar)
  • High silanol activity
  • Particle size of 2.5 μm
  • Parker-style endfitting type
  • Pore size of 100 Å and surface area of 230
  • HSS particle technology, USP Classification L43
  • Inner diameter of 3 mm and length of 150 mm
  • Carbon load of 7 %
  • One unit per package

Catalog No. 50-466-454 Supplier Waters Corp Supplier No. 186006746
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Low coverage, unendcapped pentafluorophenyl phase
  • Offers exceptional separation of closely related compounds
  • Provides alternate selectivity to traditional alkyl chain columns
  • Packed in ultra-low dispersion hardware for eXtended Performance (XP)
  • Enables robust separation performance and high throughput for HPLC assays
  • Facilitates seamless transition to UPLC adoption
  • PFP/Fluoro-Phenyl chemistry, reversed phase separation mode
  • Silica particle substrate with spherical shape
  • pH range of 2 to 8
  • Maximum pressure of 18000 psi (1240 Bar)
  • High silanol activity
  • Particle size of 2.5 μm
  • Parker-style endfitting type
  • Pore size of 100 Å and surface area of 230
  • HSS particle technology, USP Classification L43
  • Inner diameter of 3 mm and length of 150 mm
  • Carbon load of 7 %
  • One unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.