Learn More
Waters Corp XSelect HSS PFP Column, 3.5 μm, 2.1 x 75 mm, 100Å
Catalog No. 50179335 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP column provides diverse analyte selectivity, high analyte loading, and operational stability. It offers full scalability for UPLC, HPLC, and preparative separation platforms. This fluorinated phase is a useful alternative to standard C18 phases, particularly for selectivity with bases or planar aromatic compounds. Its High Strength Silica (HSS) technology ensures increased polar compound retention and mechanical strength for UPLC pressures.
  • Offers diverse analyte selectivity and high analyte loading
  • Ensures operational stability and full scalability for separation platforms
  • Provides alternatives to standard C18 phases
  • Designed for selectivity with bases or planar aromatic compounds
  • Utilizes High Strength Silica (HSS) technology
  • Achieves increased polar compound retention
  • Tolerates UPLC pressures
  • Aims for unmatched reproducibility and selectivity
Catalog No. 50-179-335 Supplier Waters Corp Supplier No. 186005848
The XSelect HSS PFP column provides diverse analyte selectivity, high analyte loading, and operational stability. It offers full scalability for UPLC, HPLC, and preparative separation platforms. This fluorinated phase is a useful alternative to standard C18 phases, particularly for selectivity with bases or planar aromatic compounds. Its High Strength Silica (HSS) technology ensures increased polar compound retention and mechanical strength for UPLC pressures.
  • Offers diverse analyte selectivity and high analyte loading
  • Ensures operational stability and full scalability for separation platforms
  • Provides alternatives to standard C18 phases
  • Designed for selectivity with bases or planar aromatic compounds
  • Utilizes High Strength Silica (HSS) technology
  • Achieves increased polar compound retention
  • Tolerates UPLC pressures
  • Aims for unmatched reproducibility and selectivity