Learn More
Waters Corp XSelect HSS PFP Column, 100Å, 3.5 µm, 3 mm X 50 mm, 1/pk
Catalog No. 50179340 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. XSelect HSS PFP HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
Catalog No. 50-179-340 Supplier Waters Corp Supplier No. 186005853
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. XSelect HSS PFP HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.