Learn More
Waters Corp XSelect HSS PFP Column, 100Å, 3.5 μm, 4.6 mm X 100 mm
Catalog No. 50179348 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP column is a non-endcapped, silica-based HPLC column designed for low pH separations. It offers alternative selectivity compared to C18 phases and provides dependable, scalable PFP performance using HSS technology for enhanced polar molecule retention and UPLC pressure resistance.
  • Non-endcapped silica-based design for low pH separations.
  • Offers alternative selectivity to C18 phases.
  • Scalable for UPLC and HPLC platforms.
  • High Strength Silica (HSS) technology ensures enhanced polar molecule retention and flexible selectivity.
  • Delivers dependable PFP performance with specialized HSS substrate and PFP ligand.
  • Ideal for improving retention of challenging polar basic analytes.
Catalog No. 50-179-348 Supplier Waters Corp Supplier No. 186005861
Add to Cart
Add to Cart
The XSelect HSS PFP column is a non-endcapped, silica-based HPLC column designed for low pH separations. It offers alternative selectivity compared to C18 phases and provides dependable, scalable PFP performance using HSS technology for enhanced polar molecule retention and UPLC pressure resistance.
  • Non-endcapped silica-based design for low pH separations.
  • Offers alternative selectivity to C18 phases.
  • Scalable for UPLC and HPLC platforms.
  • High Strength Silica (HSS) technology ensures enhanced polar molecule retention and flexible selectivity.
  • Delivers dependable PFP performance with specialized HSS substrate and PFP ligand.
  • Ideal for improving retention of challenging polar basic analytes.