Learn More
Waters Corp Waters XSelect HSS PFP Column, 3.5 μm, 4.6 x 30 mm, Fluoro-Phenyl
Catalog No. 50179345 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XSelect HSS PFP columns are designed for high-speed liquid chromatography, offering exceptional selectivity for bases or planar aromatic compounds. They use High Strength Silica (HSS) technology, providing superior mechanical strength and diverse retention characteristics.
  • Fluorinated phases offer alternatives to standard C18 phases for enhanced selectivity.
  • Designed for improved selectivity of bases or planar aromatic compounds.
  • High strength silica (HSS) technology provides unique retention characteristics.
  • HSS particles enhance polar compound retention and offer different selectivity.
  • Ensures excellent reproducibility and method transferability.
  • Seamlessly scalable across various particle sizes and column formats.
  • Features low column bleed and excellent batch-to-batch reproducibility.
Catalog No. 50-179-345 Supplier Waters Corp Supplier No. 186005858
These XSelect HSS PFP columns are designed for high-speed liquid chromatography, offering exceptional selectivity for bases or planar aromatic compounds. They use High Strength Silica (HSS) technology, providing superior mechanical strength and diverse retention characteristics.
  • Fluorinated phases offer alternatives to standard C18 phases for enhanced selectivity.
  • Designed for improved selectivity of bases or planar aromatic compounds.
  • High strength silica (HSS) technology provides unique retention characteristics.
  • HSS particles enhance polar compound retention and offer different selectivity.
  • Ensures excellent reproducibility and method transferability.
  • Seamlessly scalable across various particle sizes and column formats.
  • Features low column bleed and excellent batch-to-batch reproducibility.