Learn More
Waters Corp Waters Column, PFP, 2.5 μm, 4.6 mm ID, 150 mm Length
Supplier: Waters Corp 186006747
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase designed for exceptional separation of closely related compounds. These 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance and robustness for HPLC assays, enabling a seamless transition to UPLC.
- PFP/Fluoro-Phenyl chemistry for reversed phase separation on a silica substrate.
- Features 2.5 μm particle size and 100 Å pore size.
- Withstands a maximum pressure of 10000 psi (690 Bar); pH range of 2 to 8.
- Compatible with UPLC and UHPLC systems.
- High Strength Silica (HSS) technology for diverse selectivity and increased retention of polar molecules.
- Recommended for higher retention of basic polar analytes, leveraging unique pi-pi interactions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.