Learn More
Waters Corp Waters XSelect HSS PFP Column, 5 μm, 3 mm × 100 mm, 100 Å
Catalog No. 50179362 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. These HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
  • PFP/Fluoro-Phenyl chemistry for alternative selectivity.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • Supports a pH range of 2 to 8.
  • Withstands maximum pressure of 6000 psi (415 Bar).
  • Non-endcapped design.
  • High silanol activity.
  • Spherical particle shape.
  • HSS particle technology.
  • L43 USP classification.
  • 7% carbon load.
  • Designed for HPLC systems.
Catalog No. 50-179-362 Supplier Waters Corp Supplier No. 186005877
Add to Cart
Add to Cart
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. These HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
  • PFP/Fluoro-Phenyl chemistry for alternative selectivity.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • Supports a pH range of 2 to 8.
  • Withstands maximum pressure of 6000 psi (415 Bar).
  • Non-endcapped design.
  • High silanol activity.
  • Spherical particle shape.
  • HSS particle technology.
  • L43 USP classification.
  • 7% carbon load.
  • Designed for HPLC systems.