Learn More
Waters Corp WATERS CORPORATION XSelect HSS PFP Column, 100Å, 5 μm, 3 mm x 50 mm
Catalog No. 50179361 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. This column is fully scalable and transferable to ACQUITY UPLC separations.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Non-endcapped
  • PFP bonding technology
  • High silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 100 Å pore size
  • 230 surface area
  • HPLC system compatibility
  • HSS particle technology
  • L43 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 7% carbon load
  • 1 unit per package
Catalog No. 50-179-361 Supplier Waters Corp Supplier No. 186005875
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. This column is fully scalable and transferable to ACQUITY UPLC separations.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Non-endcapped
  • PFP bonding technology
  • High silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 100 Å pore size
  • 230 surface area
  • HPLC system compatibility
  • HSS particle technology
  • L43 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 7% carbon load
  • 1 unit per package