Learn More
Waters Corp Waters XSelect HSS PFP Column, 100Å, 5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186005885
Encompass_Preferred
This non-endcapped silica-based HPLC column is designed for low pH separations, offering alternative selectivity to C18 phases. It is fully scalable and transferable to ACQUITY UPLC. The HSS substrate and PFP ligand preserve peak shape and retention. High Strength Silica (HSS) technology provides varied selectivity and increased retention for polar molecules, with mechanical strength for UPLC pressures. Useful for higher compound retention of polar basic analytes.
  • Non-endcapped silica for low pH separations, offering alternative selectivity.
  • Scalable and transferable to ACQUITY UPLC.
  • Preserves peak shape and retention with HSS substrate and PFP ligand.
  • High Strength Silica (HSS) technology for polar molecule retention and UPLC pressure endurance.
  • Offers higher compound retention for polar basic analytes.
  • PFP phases enable pi-pi interaction and extra selectivity.
Catalog No. 50-179-370