Learn More
Waters Corp HPLC Column, XSelect HSS PFP, 5 μm, 4.6 x 50 mm, 100 Å
Catalog No. 50179367 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP column is a low-coverage pentafluorophenyl phase, offering excellent separation of closely related compounds and alternative selectivity to conventional alkyl chain columns. It is designed for reproducible and highly selective chromatographic separations, utilizing Charged Surface Hybrid (CSH) and High Strength Silica (HSS) particle technologies. This specific version has an inner diameter of 4.6 mm, a length of 50 mm, and a particle size of 5.0 μm.
  • Low-coverage pentafluorophenyl (PFP) phase
  • No end caps
  • Offers excellent separation of closely related compounds
  • Provides alternative selectivity to conventional alkyl chain columns
  • Designed for reproducible and highly selective chromatographic separations
  • Utilizes Charged Surface Hybrid (CSH) and High Strength Silica (HSS) particle technologies
Catalog No. 50-179-367 Supplier Waters Corp Supplier No. 186005882
The XSelect HSS PFP column is a low-coverage pentafluorophenyl phase, offering excellent separation of closely related compounds and alternative selectivity to conventional alkyl chain columns. It is designed for reproducible and highly selective chromatographic separations, utilizing Charged Surface Hybrid (CSH) and High Strength Silica (HSS) particle technologies. This specific version has an inner diameter of 4.6 mm, a length of 50 mm, and a particle size of 5.0 μm.
  • Low-coverage pentafluorophenyl (PFP) phase
  • No end caps
  • Offers excellent separation of closely related compounds
  • Provides alternative selectivity to conventional alkyl chain columns
  • Designed for reproducible and highly selective chromatographic separations
  • Utilizes Charged Surface Hybrid (CSH) and High Strength Silica (HSS) particle technologies