Learn More
Waters Corp Column, 100 Å, 3.5 μm, 4.6 mm, 150 mm
Catalog No. 502390963 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP Column is a non-endcapped, silica-based HPLC column for low pH separations. It is scalable to ACQUITY UPLC separations and includes an eConnect tag for automatic identification and tracking, optimized with Alliance iS System and Empower software.
  • PFP/Fluoro-phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 1 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Non-endcapped
  • PFP bonding technology
  • High silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • 4.6 mm inner diameter
  • 150 mm length
  • USP classification L43
  • Suitable for small molecule applications
  • One unit per package
  • Equipped with eConnect column tag for automatic identification, tracking, and digital tracing of column use history
Catalog No. 50-239-0963 Supplier Waters Corp Supplier No. 186005862RF
The XSelect HSS PFP Column is a non-endcapped, silica-based HPLC column for low pH separations. It is scalable to ACQUITY UPLC separations and includes an eConnect tag for automatic identification and tracking, optimized with Alliance iS System and Empower software.
  • PFP/Fluoro-phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 1 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Non-endcapped
  • PFP bonding technology
  • High silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • 4.6 mm inner diameter
  • 150 mm length
  • USP classification L43
  • Suitable for small molecule applications
  • One unit per package
  • Equipped with eConnect column tag for automatic identification, tracking, and digital tracing of column use history