Learn More
Waters Corp XSelect HSS PFP Method Validation Kit, 100 Å, 3.5 μm, 2.1 mm x 100 mm
Catalog No. 502392379 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP Method Validation Kit is designed for method validation, featuring PFP/Fluoro-Phenyl chemistry and HSS particle technology. It provides a comprehensive solution for analytical needs.
  • Designed for method validation
  • Features PFP/Fluoro-Phenyl chemistry
  • Utilizes HSS particle technology
  • Includes columns with 100 Å pore size
  • Comes with 3.5 μm particle size
  • Offers 2.1 mm inner diameter
  • Has a 100 mm length
  • Contains 3 units per package
Catalog No. 50-239-2379 Supplier Waters Corp Supplier No. 186005890
The XSelect HSS PFP Method Validation Kit is designed for method validation, featuring PFP/Fluoro-Phenyl chemistry and HSS particle technology. It provides a comprehensive solution for analytical needs.
  • Designed for method validation
  • Features PFP/Fluoro-Phenyl chemistry
  • Utilizes HSS particle technology
  • Includes columns with 100 Å pore size
  • Comes with 3.5 μm particle size
  • Offers 2.1 mm inner diameter
  • Has a 100 mm length
  • Contains 3 units per package